Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | 99.8% | 252.31 g·mol⁻¹ | C20H12 | 50 MG
SDP

Catalog No. 5000241532
Encompass_Preferred

Benzo[a]pyrene (Standard) is an analytical reference standard of the polycyclic aromatic hydrocarbon benzo[a]pyrene, provided as a high-purity solid for research and analytical use. It is suitable for chromatographic calibration, method development, and toxicology studies.

  • Analytical standard for quantitative and qualitative assays.
  • High purity (≈99.8%).
  • Molecular formula C20H12 and molecular weight 252.31 g·mol⁻¹.
  • Available in small vials (5 mg-100 mg; larger sizes by quote).
  • Intended for chromatographic calibration, method development, and chemoprevention research.
  • CAS number 50-32-8 for unambiguous identification.

Catalog No. 50-002-41532 Supplier Medchemexpress LLC Supplier No. HY107377R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzo[a]pyrene (Standard) is an analytical reference standard of the polycyclic aromatic hydrocarbon benzo[a]pyrene, provided as a high-purity solid for research and analytical use. It is suitable for chromatographic calibration, method development, and toxicology studies.

  • Analytical standard for quantitative and qualitative assays.
  • High purity (≈99.8%).
  • Molecular formula C20H12 and molecular weight 252.31 g·mol⁻¹.
  • Available in small vials (5 mg-100 mg; larger sizes by quote).
  • Intended for chromatographic calibration, method development, and chemoprevention research.
  • CAS number 50-32-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.