Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene, 96%

Catalog No. AA1585602
Change view
Click to view available options
Quantity:
100 mg
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1585602 0.5 g
AA15856MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1585602 Supplier Thermo Scientific Chemicals Supplier No. 01585602
Only null left
Add to Cart

Extensively used in cancer research | CAS: 50-32-8 | C20H12 | 252.32 g/mol

Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name benzo[a]pyrene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

Melting Point 177°C to 180°C
Boiling Point 495°C
Quantity 0.5 g
UN Number UN3077
Merck Index 14,1103
Solubility Information Soluble in benzene, toluene, and xylene; Sparingly soluble in alcohol, methanol; Insoluble in water
Formula Weight 252.3
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Benzo[a]pyrene
Hazard Category H317-H340-H350-H360FD
Hazard Statement GHS H Statement
H340-H350-H360-H317
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-028-5
RTECSNumber DJ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.