Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene 98%, Thermo Scientific™

Catalog No. AC105601000
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105601000 100 mg
AC105600010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105601000 Supplier Thermo Scientific Chemicals Supplier No. 105601000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name pentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 175°C to 179°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 100 mg
Beilstein 05, IV, 2687
Merck Index 15, 1106
Formula Weight 252.31
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Benzo[a]pyrene, 98%
Show More Show Less
EINECSNumber 200-028-5
RTECSNumber DJ3675000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.