Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019054 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9054 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9054 Supplier MilliporeSigma Supelco Supplier No. 5196850MG
Add to Cart
Add to Cart

Benzo[a]pyrene is a high molecular weight, polycyclic aromatic hydrocarbon, which is formed the pyrolysis of organic matter, and are present in the environment particularly in the atmosphere as particulate matter.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 50-32-8
Product Line TraceCERT
Physical Form Neat
Linear Formula C20H12
MDL Number MFCD00003602
Melting Point 177°C to 180°C
Boiling Point 495°C
Synonym 3,4-Benzopyrene; 3,4-Benzpyrene; Benzo[def]chrysene
Quantity 50 mg
Formula Weight 252.31
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.