Learn More
Medchemexpress LLC Benzo[a]pyrene-d12 | 63466-71-7 | MFCD00143664 | 99.3% | 264.38 | C20D12 | 5 MG
SDP
Catalog No. 5000240865
Encompass_Preferred
Benzo[a]pyrene-d12 is the deuterium-labeled analogue of benzo[a]pyrene supplied as a high-purity stable isotope standard for analytical and research use. It functions as an internal standard or tracer for mass spectrometry and quantitative studies in environmental analysis, toxicology, and chemoprevention research.
  • Deuterium-labeled analogue for mass spectrometry tracing and quantification.
  • High purity (99.3%) for reliable analytical performance.
  • Available in small-mass pack sizes suitable for standards preparation.
  • Molecular formula C20D12 and molecular weight 264.38 for exact mass calculations.
  • Stable under recommended storage conditions with extended shelf life.
Catalog No. 50-002-40865 Supplier Medchemexpress LLC Supplier No. HY107377S5MG
Benzo[a]pyrene-d12 is the deuterium-labeled analogue of benzo[a]pyrene supplied as a high-purity stable isotope standard for analytical and research use. It functions as an internal standard or tracer for mass spectrometry and quantitative studies in environmental analysis, toxicology, and chemoprevention research.
  • Deuterium-labeled analogue for mass spectrometry tracing and quantification.
  • High purity (99.3%) for reliable analytical performance.
  • Available in small-mass pack sizes suitable for standards preparation.
  • Molecular formula C20D12 and molecular weight 264.38 for exact mass calculations.
  • Stable under recommended storage conditions with extended shelf life.