Learn More
Medchemexpress LLC Benzo[a]pyrene-d12 | 63466-71-7 | MFCD00143664 | 99.3% | 264.38 g/mol | C20D12 | 10 MG
SDP
Catalog No. 5000240647
Encompass_Preferred
Benzo[a]pyrene-d12 is the deuterium-labeled isotopologue of benzo[a]pyrene supplied as a research-use analytical standard. It is used as an internal standard or tracer in studies of benzo[a]pyrene metabolism, quantification of polycyclic aromatic hydrocarbons (PAHs), and chemoprevention research. The isotopic label improves accuracy and precision in mass-spectrometry based assays.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic and chemical purity suitable for trace-level measurements.
  • Compatible with LC-MS and GC-MS workflows.
  • Useful as a tracer in metabolism and chemoprevention studies.
  • Supplied in small, accurately weighed quantities for analytical use.
Catalog No. 50-002-40647 Supplier Medchemexpress LLC Supplier No. HY107377S10MG
Benzo[a]pyrene-d12 is the deuterium-labeled isotopologue of benzo[a]pyrene supplied as a research-use analytical standard. It is used as an internal standard or tracer in studies of benzo[a]pyrene metabolism, quantification of polycyclic aromatic hydrocarbons (PAHs), and chemoprevention research. The isotopic label improves accuracy and precision in mass-spectrometry based assays.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic and chemical purity suitable for trace-level measurements.
  • Compatible with LC-MS and GC-MS workflows.
  • Useful as a tracer in metabolism and chemoprevention studies.
  • Supplied in small, accurately weighed quantities for analytical use.