Learn More
Medchemexpress LLC Benzo[a]pyrene-d12 | 63466-71-7 | MFCD00143664 | 99.3% | 264.4 g/mol | C20D12 | 1 MG
SDP
Catalog No. 5000240646
Encompass_Preferred
Benzo[a]pyrene-d12 is the deuterium-labeled analog of benzo[a]pyrene, provided as a high-purity stable isotope standard for research and analytical workflows. It is intended for use as an internal standard in mass spectrometry, quantitation of polycyclic aromatic hydrocarbons, and studies of carcinogenicity and chemoprevention.
  • High isotopic enrichment and purity (99.3%).
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Useful for environmental, toxicology, and chemoprevention research.
  • Chemical formula C20D12; molecular weight 264.4 g/mol.
  • Supplied as a 1 mg vial for analytical applications.
Catalog No. 50-002-40646 Supplier Medchemexpress LLC Supplier No. HY107377S1MG
Benzo[a]pyrene-d12 is the deuterium-labeled analog of benzo[a]pyrene, provided as a high-purity stable isotope standard for research and analytical workflows. It is intended for use as an internal standard in mass spectrometry, quantitation of polycyclic aromatic hydrocarbons, and studies of carcinogenicity and chemoprevention.
  • High isotopic enrichment and purity (99.3%).
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Useful for environmental, toxicology, and chemoprevention research.
  • Chemical formula C20D12; molecular weight 264.4 g/mol.
  • Supplied as a 1 mg vial for analytical applications.