Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene, For environmental analysis, MilliporeSigma™ Supelco™

Catalog No. 111018470 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8470 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8470 Supplier MilliporeSigma Supelco Supplier No. 48564
Add to Cart
Add to Cart

Benzo[a]pyrene belongs to the class of polycyclic aromatic hydrocarbons, which can exist as a potent genotoxic compound majorly formed via pyrolysis of organic matter and can also be found in industrial and cigarette smoke, coal tar and food.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-32-8
Physical Form Neat
Linear Formula C20H12
MDL Number MFCD00003602
Melting Point 177°C to 180°C
Boiling Point 495°C
Synonym 3,4-Benzopyrene; 3,4-Benzpyrene; Benzo[def]chrysene
Type Organic Pollutant Standard
Industry Type Environmental Testing
Quantity 100 mg
Formula Weight 252.31
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.