Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene Solution, 100 μg/mL in cycloHexane, MilliporeSigma™ Supelco™

Catalog No. 111018932 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8932 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8932 Supplier MilliporeSigma Supelco Supplier No. 3130610ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzo[a]pyrene belongs to the family of polycyclic aromatic hydrocarbon (PAH) compounds, which consist of two or more aromatic rings made up of only hydrogen and carbon elements.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzo[a]pyrene
CAS 50-32-8, 110-82-7, 50-32-8
Flash Point -18°C
Concentration 100 μg/mL
Linear Formula C20H12
Technique Gas Chromatography
MDL Number MFCD00003602
Type Analytical Standard
Solvent or Matrix Cyclohexane
Industry Type Environmental Testing
Molecular Weight (g/mol) 252.31
Quantity 10 mL
Formula Weight 252.31
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.