Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzo[a]pyrene (standard) | 50-32-8 | MFCD00003602 | 99.8% | 252.31 | C20H12 | 10 MG
SDP

Catalog No. 5000241386
Encompass_Preferred

Benzo[a]pyrene is an analytical reference standard intended for research and analytical applications, commonly used for method development, calibration, and studies of chemoprevention and carcinogenicity. The material is supplied as a high-purity solid in small laboratory pack sizes suitable for analytical workflows.

  • Purity 99.8%.
  • Molecular weight 252.31 g/mol.
  • Chemical formula C20H12.
  • Appearance light yellow to green yellow solid.
  • Available in small pack sizes appropriate for analytical use.
  • Store sealed, away from light and moisture; follow certificate of analysis for conditions.

Catalog No. 50-002-41386 Supplier Medchemexpress LLC Supplier No. HY107377R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzo[a]pyrene is an analytical reference standard intended for research and analytical applications, commonly used for method development, calibration, and studies of chemoprevention and carcinogenicity. The material is supplied as a high-purity solid in small laboratory pack sizes suitable for analytical workflows.

  • Purity 99.8%.
  • Molecular weight 252.31 g/mol.
  • Chemical formula C20H12.
  • Appearance light yellow to green yellow solid.
  • Available in small pack sizes appropriate for analytical use.
  • Store sealed, away from light and moisture; follow certificate of analysis for conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.