Learn More
Medchemexpress LLC Benzo[a]pyrene (standard) | 50-32-8 | MFCD00003602 | 99.8% | 252.31 g·mol-1 | C20H12 | 100 MG
SDP
Catalog No. 5000241475
Encompass_Preferred
Benzo[a]pyrene (Standard) is an analytical reference standard of the polycyclic aromatic hydrocarbon benzo[a]pyrene (CAS 50-32-8). It is supplied as a high-purity solid for qualitative and quantitative analysis and is intended for method development, calibration, and toxicology research.
  • High purity analytical standard (99.8%).
  • Molecular formula C20H12, molecular weight 252.31 g·mol-1.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Supplied as a solid packaged at 100 mg.
  • Includes safety data sheet documentation for handling and storage.
Catalog No. 50-002-41475 Supplier Medchemexpress LLC Supplier No. HY107377R100MG
Benzo[a]pyrene (Standard) is an analytical reference standard of the polycyclic aromatic hydrocarbon benzo[a]pyrene (CAS 50-32-8). It is supplied as a high-purity solid for qualitative and quantitative analysis and is intended for method development, calibration, and toxicology research.
  • High purity analytical standard (99.8%).
  • Molecular formula C20H12, molecular weight 252.31 g·mol-1.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Supplied as a solid packaged at 100 mg.
  • Includes safety data sheet documentation for handling and storage.