Learn More
Crescent Chemical Co Inc BENZO(B)FLUORANTHENE D12 1ML
SDP
Catalog No. 5079611
Encompass_Preferred
Benzo(b)fluoranthene D12 10 µg/mL in Cyclohexane Polycyc.arom.hydrocarb (Isotop Lsg) CAS 93951-98-5 Molecular Weight - 264.3832 Storage - RT Catalog Num - DRE-LA20565100CY chemical
Catalog No. 50-796-11 Supplier Crescent Chemical Co Inc Supplier No. PSLA20565100CY
Benzo(b)fluoranthene D12 10 µg/mL in Cyclohexane Polycyc.arom.hydrocarb (Isotop Lsg) CAS 93951-98-5 Molecular Weight - 264.3832 Storage - RT Catalog Num - DRE-LA20565100CY chemical