Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]furan-2,5-diboronic acid, 98%, Thermo Scientific™

Catalog No. AAH53025MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53025MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53025MD Supplier Thermo Scientific Chemicals Supplier No. H53025MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-69-5
Molecular Formula C8H8B2O5
Molecular Weight (g/mol) 205.767
MDL Number MFCD08436058
InChI Key LCHGNXMXVDSVAR-UHFFFAOYSA-N
Synonym benzofuran-2,5-diboronic acid, benzofuran-2,5-diyldiboronic acid, benzofuran-2,5-yldiboronic acid, boronic acid, 2,5-benzofurandiylbis-9ci, 5-dihydroxyboranyl-1-benzofuran-2-ylboronic acid, acmc-209r9u, benzo b furan-2,5-diboronic acid, 1-benzofuran-2,5-diyldiboronic acid
PubChem CID 44119346
IUPAC Name (2-borono-1-benzofuran-5-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)OC(=C2)B(O)O)(O)O

Specifications

Melting Point 293°C to 295°C
Quantity 250 mg
Formula Weight 205.77
Percent Purity 98%
Chemical Name or Material Benzo[b]furan-2,5-diboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.