Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]naphtho[2,3-d]thiophene 99.5+%, Thermo Scientific™

Catalog No. AC290942500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290942500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290942500 Supplier Thermo Scientific Chemicals Supplier No. 290942500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243-46-9
Molecular Formula C16H10S
Molecular Weight (g/mol) 234.316
InChI Key UWMISBRPSJFHIR-UHFFFAOYSA-N
Synonym benzo b naphtho 2,3-d thiophene, naphtho 2,3-b 1 benzothiophene, 17-thiatetracyclo 8.7.0.0 3 ,?.0 1 1 , 1 ? heptadeca-1 10 ,2,4,6,8,11 16 ,12,14-octaene, naphtho 2,3-b 1 benzothiole, naphtho 2,3-b benzo b thiophene, naphtho 2,3-b 1 benzothiophene #, benzo b naphtho 2,3-d thiophene, bcr r certified reference material
PubChem CID 9203
IUPAC Name naphtho[2,3-b][1]benzothiole
SMILES C1=CC=C2C=C3C(=CC2=C1)C4=CC=CC=C4S3

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 159°C to 161°C
Color White
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 234.32
Percent Purity ≥99.50%
Physical Form Powder
Chemical Name or Material Benzo[b]naphtho[2, 3-d]thiophene, 99.5+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.