Learn More
Medchemexpress LLC Benzo[b]thiophene-2-carboxylic acid, 5,6-diethoxy-, sodium salt (1:1) | 105102-18-9 | MFCD00864802 | 99.7% | 288.3 g/mol | C13H13O4S.Na | 50 MG
SDP
Catalog No. 5000245574
Encompass_Preferred
Tibenelast sodium is the sodium salt form of a benzo[b]thiophene carboxylic acid derivative that functions as a phosphodiesterase (PDE) inhibitor. It is provided as a research-grade small molecule for use in pharmacology and inflammation/respiratory studies, supplied as a solid powder for laboratory experiments.
  • High purity (≈99.7%) suitable for research use.
  • Phosphodiesterase inhibitor activity for inflammation and respiratory research.
  • Sodium salt form for enhanced solubility in polar solvents like DMSO.
  • Solid powder presentation convenient for storage and formulation.
  • Identified by CAS 105102-18-9 for unambiguous referencing.
Catalog No. 50-002-45574 Supplier Medchemexpress LLC Supplier No. HY10170550MG
Tibenelast sodium is the sodium salt form of a benzo[b]thiophene carboxylic acid derivative that functions as a phosphodiesterase (PDE) inhibitor. It is provided as a research-grade small molecule for use in pharmacology and inflammation/respiratory studies, supplied as a solid powder for laboratory experiments.
  • High purity (≈99.7%) suitable for research use.
  • Phosphodiesterase inhibitor activity for inflammation and respiratory research.
  • Sodium salt form for enhanced solubility in polar solvents like DMSO.
  • Solid powder presentation convenient for storage and formulation.
  • Identified by CAS 105102-18-9 for unambiguous referencing.