Learn More
Medchemexpress LLC Benzo[b]thiophene-2-carboxylic acid, 5,6-diethoxy-, sodium salt | 105102-18-9 | MFCD00864802 | >98.0% | 288.30 g/mol | C13H13NaO4S | 100 MG
SDP
Catalog No. 5000249131
Encompass_Preferred
Tibenelast sodium is the sodium salt of benzo[b]thiophene-2-carboxylic acid, 5,6-diethoxy- (synonym LY-186655), a small-molecule phosphodiesterase inhibitor supplied for laboratory research use. It is identified by CAS 105102-18-9 and has a molecular weight of 288.30 g/mol.
  • Phosphodiesterase inhibitor activity useful for biochemical and pharmacology studies.
  • Provided as a research reagent for laboratory use only, not for human or veterinary applications.
  • Reported purity greater than 98.0%.
  • Small-package quantities suitable for early-stage research, such as 25-100 mg.
  • Chemical formula C13H13NaO4S and molecular weight 288.30 g/mol.
Catalog No. 50-002-49131 Supplier Medchemexpress LLC Supplier No. HY101705100MG
Tibenelast sodium is the sodium salt of benzo[b]thiophene-2-carboxylic acid, 5,6-diethoxy- (synonym LY-186655), a small-molecule phosphodiesterase inhibitor supplied for laboratory research use. It is identified by CAS 105102-18-9 and has a molecular weight of 288.30 g/mol.
  • Phosphodiesterase inhibitor activity useful for biochemical and pharmacology studies.
  • Provided as a research reagent for laboratory use only, not for human or veterinary applications.
  • Reported purity greater than 98.0%.
  • Small-package quantities suitable for early-stage research, such as 25-100 mg.
  • Chemical formula C13H13NaO4S and molecular weight 288.30 g/mol.