Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]thiophene-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B47745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4774-5G 5 g
B47741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4774-5G Supplier TCI America Supplier No. B47745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 113893-08-6
Molecular Formula C8H7BO2S
Molecular Weight (g/mol) 178.012
MDL Number MFCD01114669
InChI Key QVANIYYVZZLQJP-UHFFFAOYSA-N
PubChem CID 2776344
IUPAC Name 1-benzothiophen-3-ylboronic acid
SMILES B(C1=CSC2=CC=CC=C12)(O)O

Specifications

Melting Point 230°C
Quantity 5 g
Synonym benzothiophene-3-boronic acid, benzo b thiophen-3-ylboronic acid, benzo b thien-3-ylboronic acid, benzo b thiophene-3-boronic acid, thianaphthene-3-boronic acid
Formula Weight 178.01
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.