Learn More
Medchemexpress LLC Benzo[b]thiophene-3-carbaldehyde | 5381-20-4 | 162.21 | 10 G
SDP
Catalog No. 5000445588
Encompass_Preferred
Benzo[b]thiophene-3-carbaldehyde is a 3-formyl-substituted benzo[b]thiophene derivative that functions as an oxidizable substrate, converting to benzo[b]thiophene-3-carboxylic acid under liquid-phase conditions. It exhibits lower oxidation difficulty compared to similar compounds and its reaction rate can be significantly increased in the presence of initiators. This compound also serves as a crucial starting material for synthesizing other compounds, including heteroaryl chalcones.
  • Acts as an oxidizable substrate
  • Converts to benzo[b]thiophene-3-carboxylic acid
  • Exhibits lower oxidation difficulty
  • Reaction rate is significantly increased by initiators (e.g., Na+Br- or DBA)
  • Crucial starting material for synthesizing heteroaryl chalcones
  • In vitro, oxidation to benzo[b]thiophene-3-carboxylic acid occurs within 60 hours without initiator, or 6 hours with initiators.
Catalog No. 50-004-45588 Supplier Medchemexpress LLC Supplier No. HYW00225110G
Benzo[b]thiophene-3-carbaldehyde is a 3-formyl-substituted benzo[b]thiophene derivative that functions as an oxidizable substrate, converting to benzo[b]thiophene-3-carboxylic acid under liquid-phase conditions. It exhibits lower oxidation difficulty compared to similar compounds and its reaction rate can be significantly increased in the presence of initiators. This compound also serves as a crucial starting material for synthesizing other compounds, including heteroaryl chalcones.
  • Acts as an oxidizable substrate
  • Converts to benzo[b]thiophene-3-carboxylic acid
  • Exhibits lower oxidation difficulty
  • Reaction rate is significantly increased by initiators (e.g., Na+Br- or DBA)
  • Crucial starting material for synthesizing heteroaryl chalcones
  • In vitro, oxidation to benzo[b]thiophene-3-carboxylic acid occurs within 60 hours without initiator, or 6 hours with initiators.