Learn More
Medchemexpress LLC Benzo[b]thiophene-3-carbonitrile, 2-amino-4-[(4aS)-8-chloro-10-fluoro-2,3,4,4a,5,6-hexahydro-12... | 2649788-46-3 | 98.8% | 528.96 | 1 MG
SDP
Catalog No. 5000391348
Encompass_Preferred
KRAS G12C inhibitor 67 is provided as a white to off-white solid with a high purity of 98.80%. This product has been tested and complies with the given specifications. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 98.8% (LCMS)
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
Catalog No. 50-003-91348 Supplier Medchemexpress LLC Supplier No. HY1643151MG
KRAS G12C inhibitor 67 is provided as a white to off-white solid with a high purity of 98.80%. This product has been tested and complies with the given specifications. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 98.8% (LCMS)
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month