Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzo[b]thiophene, 98+%

Manufacturer:  Alfa Aesar™ A1232806

Quantity 5g

 View more versions of this product

Catalog No. AAA1232806


  Check Availability
Add to cart

Specifications

Description

Applications
Benzo[b]thiophene is used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium. It is used in organic as well as pharmaceutical industry as a component in the synthesis of raloxifene, a breast cancer therapeutic, or hydrodesulfurization reactions.

Solubility
Soluble in water (0.13 g/L at 20°C), chloroform and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

95-15-8
134.2
UN3077
14,9303
Benzo[b]thiophene
C8H6S
MFCD00005864
80580
1-Benzothiophene; Thianaphthene

Specifications

95-15-8
134.2
UN3077
14,9303
221° to 222°C
5g
Unpleasant
≥98%
C8H6S
MFCD00005864
80580
1-Benzothiophene; Thianaphthene
28° to 32°C
91°C (195°F)
Benzo[b]thiophene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P273
Wear protective clothing.
Avoid release to the environment.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit