Learn More
Medchemexpress LLC Benzo[f]quinoxaline-7-sulfonamide, 1,2,3,4-tetrahydro-6-nitro-2,3-dioxo-, sodium salt (1:2) | 479347-86-9 | C12H6N4Na2O6S | 25 MG
SDP
Catalog No. 5000374467
Encompass_Preferred
NBQX disodium is a highly selective and competitive antagonist of the AMPA receptor, exhibiting neuroprotective and anticonvulsant activity. It possesses a high affinity for AMPA and kainate binding sites, with minimal affinity for the glutamate recognition site on the NMDA receptor complex.
  • Highly selective and competitive AMPA receptor antagonist
  • Exhibits neuroprotective activity
  • Shows anticonvulsant activity
  • High affinity for AMPA and kainate binding sites
  • Little to no affinity for the NMDA receptor complex
Catalog No. 50-003-74467 Supplier Medchemexpress LLC Supplier No. 25MGHY15068A
NBQX disodium is a highly selective and competitive antagonist of the AMPA receptor, exhibiting neuroprotective and anticonvulsant activity. It possesses a high affinity for AMPA and kainate binding sites, with minimal affinity for the glutamate recognition site on the NMDA receptor complex.
  • Highly selective and competitive AMPA receptor antagonist
  • Exhibits neuroprotective activity
  • Shows anticonvulsant activity
  • High affinity for AMPA and kainate binding sites
  • Little to no affinity for the NMDA receptor complex