Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[g]chrysene, 98%, Thermo Scientific™

Catalog No. AC460270010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460270010 Supplier Thermo Scientific Chemicals Supplier No. 460270010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 196-78-1
Molecular Formula C22H14
Molecular Weight (g/mol) 278.35
InChI Key JZOIZKBKSZMVRV-UHFFFAOYSA-N
Synonym benzo g chrysene, benzo a triphenylene, naphthophenanthrene, unii-e7mq64xu75, 1,2,3,4-dibenzphenanthrene, 1,2,3,4-dibenzophenanthrene, 1,2:3,4:7,8-tribenznaphthalene, benzochrysene, acmc-20gx0y
PubChem CID 9140
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=CC=CC=C4C5=CC=CC=C35

Specifications

Melting Point 126.0°C
Boiling Point 525.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 278.35
Percent Purity 98%
Chemical Name or Material Benzo[g]chrysene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.