Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[ghi]perylene, 98+%

Catalog No. AC105550050
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105550050 5 mg Glass bottle
AC105550250 25 mg Glass bottle
AC105551000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105550050 Supplier Thermo Scientific Chemicals Supplier No. 105550050
Add to Cart

CAS: 191-24-2 | C22H12 | 276.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191-24-2
Molecular Formula C22H12
Molecular Weight (g/mol) 276.33
MDL Number MFCD00004135
InChI Key GYFAGKUZYNFMBN-UHFFFAOYSA-N
Synonym benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784
PubChem CID 9117
ChEBI CHEBI:34568
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 276.0°C to 280.0°C
Color Yellow
Boiling Point >500.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 mg
Solubility Information Solubility in water: insoluble
Formula Weight 276.33
Percent Purity 98+%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material Benzo[ghi]perylene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 205-883-8
RTECSNumber DI6200500

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.