Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[ghi]perylene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018262 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8262 10 mg
11-101-7934 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8262 Supplier MilliporeSigma Supelco Supplier No. 5548810MG
Add to Cart
Add to Cart

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

TRUSTED_SUSTAINABILITY

Specifications

CAS 191-24-2
Product Line TraceCERT
Physical Form Neat
Linear Formula C22H12
MDL Number MFCD00004135
Melting Point 277°C to 279°C
Boiling Point >500°C
Synonym 1,12-Benzoperylene
Quantity 10 mg
Formula Weight 276.33
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.