Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[ghi]perylene, MilliporeSigma™ Supelco™

Catalog No. 111019371 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9371 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9371 Supplier MilliporeSigma Supelco Supplier No. 48491
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 191-24-2
Physical Form Neat
Linear Formula C22H12
MDL Number MFCD00004135
Melting Point 277°C to 279°C
Boiling Point >500°C
Synonym 1,12-Benzoperylene
Type Organic Pollutant Standard
Quantity 25 mg
Formula Weight 276.33
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.