Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[ghi]perylene 97.0+%, TCI America™
SDP

Catalog No. B2983100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2983100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2983100MG Supplier TCI America Supplier No. B2983100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 191-24-2
Molecular Formula C22H12
Molecular Weight (g/mol) 276.338
MDL Number MFCD00004135
InChI Key GYFAGKUZYNFMBN-UHFFFAOYSA-N
Synonym benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784
PubChem CID 9117
ChEBI CHEBI:34568
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2

Specifications

Melting Point 273°C
Color Yellow
Boiling Point 542°C
Quantity 100 mg
UN Number 3077
Formula Weight 276.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[ghi]perylene
RTECSNumber DI6200500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.