Learn More
Chem Service Inc Benzo(k)fluoranthene (d12) Solution, 93952-01-3, MFCD00143674, 1.2 mL
SDP
Catalog No. NC1522615
Encompass_Preferred
Molecular Formula: C20 D12, Molecular Weight: 264.388, Concentration and Solvent: 200ug/ml in Isooctane.
Catalog No. NC1522615 Supplier Chem Service Inc Supplier No. SFD75S1.2ML
Molecular Formula: C20 D12, Molecular Weight: 264.388, Concentration and Solvent: 200ug/ml in Isooctane.