Learn More
Medchemexpress LLC Benzo[k]fluoranthene | 207-08-9 | 252.31 | 1 MG
SDP
Catalog No. 5000400457
Encompass_Preferred
Benzo[k]fluoranthene is a polycyclic aromatic hydrocarbon (PAH) that possesses carcinogenic and genotoxic properties.
  • Product type: Polycyclic aromatic hydrocarbon (PAH).
  • Biological activity: Carcinogenic and genotoxic.
  • Usage: For research use only; not sold to patients.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Solution, -20°C, protected from light, with a shelf life of 2 years.
Catalog No. 50-004-00457 Supplier Medchemexpress LLC Supplier No. HY234981MG
Benzo[k]fluoranthene is a polycyclic aromatic hydrocarbon (PAH) that possesses carcinogenic and genotoxic properties.
  • Product type: Polycyclic aromatic hydrocarbon (PAH).
  • Biological activity: Carcinogenic and genotoxic.
  • Usage: For research use only; not sold to patients.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Solution, -20°C, protected from light, with a shelf life of 2 years.