Learn More
Benzo(k)fluoranthene Solution, 100 μg/mL in methylene chloride, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| CAS | 207-08-9 |
| Linear Formula | C20H12 |
| MDL Number | MFCD00046287 |
| Synonym | Benzo[k]fluoranthene solution |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 252.31 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in methylene chloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.