Learn More
Medchemexpress LLC Benzobicyclon | 156963-66-5 | 98.0% | 446.97 g/mol | C22H19ClO4S2 | 50 MG
SDP
Catalog No. 5000226652
Encompass_Preferred
Benzobicyclon analytical standard for research and analytical applications. It is a 4-HPPD inhibitor that hydrolyzes to benzobicycline in water and is used as a reference for herbicidal activity, metabolism, and environmental fate studies. Supplied as a 50 mg standard with reported purity around 98.0%.
  • High purity analytical standard suitable for reference and calibration.
  • Targets 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) activity.
  • Useful for herbicide metabolism and environmental fate studies.
  • Solid form suitable for accurate weighing and storage.
  • Supplied in a compact 50 mg quantity for analytical workflows.
Catalog No. 50-002-26652 Supplier Medchemexpress LLC Supplier No. HY11874250MG
Benzobicyclon analytical standard for research and analytical applications. It is a 4-HPPD inhibitor that hydrolyzes to benzobicycline in water and is used as a reference for herbicidal activity, metabolism, and environmental fate studies. Supplied as a 50 mg standard with reported purity around 98.0%.
  • High purity analytical standard suitable for reference and calibration.
  • Targets 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) activity.
  • Useful for herbicide metabolism and environmental fate studies.
  • Solid form suitable for accurate weighing and storage.
  • Supplied in a compact 50 mg quantity for analytical workflows.