Learn More
Medchemexpress LLC Benzobicyclon | 156963-66-5 | MFCD09751259 | 98.0% | 446.97 | C22H19ClO4S2 | 10 MG
SDP
Catalog No. 5000227346
Encompass_Preferred
Benzobicyclon analytical standard is supplied for research and analytical applications as a reference material. It is a 4-HPPD inhibitor herbicide that hydrolyzes in water to form the active herbicidal species, producing bleaching and death in susceptible plants. The material is provided at high purity for accurate analytical and testing use.
  • Intended for research and analytical applications.
  • High purity suitable for reference and analytical workflows.
  • Hydrolyzes in water to form the active herbicide.
  • Effective against grass, sedge, and broadleaf weeds.
  • Suitable for analytical quantification and method development.
Catalog No. 50-002-27346 Supplier Medchemexpress LLC Supplier No. HY11874210MG
Benzobicyclon analytical standard is supplied for research and analytical applications as a reference material. It is a 4-HPPD inhibitor herbicide that hydrolyzes in water to form the active herbicidal species, producing bleaching and death in susceptible plants. The material is provided at high purity for accurate analytical and testing use.
  • Intended for research and analytical applications.
  • High purity suitable for reference and analytical workflows.
  • Hydrolyzes in water to form the active herbicide.
  • Effective against grass, sedge, and broadleaf weeds.
  • Suitable for analytical quantification and method development.