Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzobicyclon | 156963-66-5 | MFCD09751259 | 98.0% | 446.97 g/mol | C22H19ClO4S2 | 100 MG
SDP

Catalog No. 5000228466
Encompass_Preferred

Benzobicyclon is a research-grade herbicide compound used as a 4-HPPD inhibitor and supplied as an analytical standard for laboratory and analytical applications. It is documented with CAS and molecular identity and is typically provided in a quantified solid form for testing and reference purposes.

  • Acts as a 4-HPPD (4-hydroxyphenylpyruvate dioxygenase) inhibitor.
  • Intended for research and analytical applications.
  • CAS number 156963-66-5 for unambiguous identification.
  • Molecular formula C22H19ClO4S2 and molecular weight 446.97 g/mol.
  • Supplied as a solid standard with typical purity of 98%.

Catalog No. 50-002-28466 Supplier Medchemexpress LLC Supplier No. HY118742100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzobicyclon is a research-grade herbicide compound used as a 4-HPPD inhibitor and supplied as an analytical standard for laboratory and analytical applications. It is documented with CAS and molecular identity and is typically provided in a quantified solid form for testing and reference purposes.

  • Acts as a 4-HPPD (4-hydroxyphenylpyruvate dioxygenase) inhibitor.
  • Intended for research and analytical applications.
  • CAS number 156963-66-5 for unambiguous identification.
  • Molecular formula C22H19ClO4S2 and molecular weight 446.97 g/mol.
  • Supplied as a solid standard with typical purity of 98%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.