Learn More
Medchemexpress LLC HY-118742 5mg Medchemexpress, Benzobicyclon CAS:156963-66-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1187425MG
Encompass_Preferred
Medchemexpress, HY-118742 5mg Benzobicyclon CAS:156963-66-5 Benzobicyclon is a pro-herbicide, which acts as an inhibitor of 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) in plant, and leads to bleaching and death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9510