Learn More
Medchemexpress LLC HY-Y0258 500mg Medchemexpress, Benzocaine CAS:94-09-7 Purity:>98%
SDP
Catalog No. 501970758
Encompass_Preferred
Medchemexpress, HY-Y0258 500mg Benzocaine CAS:94-09-7 Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na+ channel, with an IC50 of 0.8 mM tested with a potential of +30 mV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0758 Supplier Medchemexpress LLC Supplier No. HYY0258500MG
Medchemexpress, HY-Y0258 500mg Benzocaine CAS:94-09-7 Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na+ channel, with an IC50 of 0.8 mM tested with a potential of +30 mV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.