Learn More
Apexbio Technology LLC Benzocaine 94-09-7 5g
SDP
Catalog No. 502240690
Encompass_Preferred
Benzocaine (94-09-7) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to inhibit these channels thereby suppressing nerve impulse conduction and local nociception Benzocaine exerts its biological activity primarily by blocking voltage-gated sodium channels and reducing sodium influx In electrophysiological assays benzocaine demonstrates inhibitory activity with an IC50 value of approximately 0 8 1 5 mM depending on assay conditions and cell models Based on these pharmacological properties benzocaine holds research potential in studies of pain sensation nerve signaling and topical anesthetic mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0690 Supplier Apexbio Technology LLC Supplier No. B16755000
Benzocaine (94-09-7) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to inhibit these channels thereby suppressing nerve impulse conduction and local nociception Benzocaine exerts its biological activity primarily by blocking voltage-gated sodium channels and reducing sodium influx In electrophysiological assays benzocaine demonstrates inhibitory activity with an IC50 value of approximately 0 8 1 5 mM depending on assay conditions and cell models Based on these pharmacological properties benzocaine holds research potential in studies of pain sensation nerve signaling and topical anesthetic mechanisms