Learn More
Medchemexpress LLC Benzocaine-d4 | 342611-08-9 | 169.21 g/mol | C9H7D4NO2 | 5mg
SDP
Catalog No. 5000267357
Encompass_Preferred
Benzocaine-d4 is the deuterium labeled Benzocaine Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na channel with an IC50 of 0 8 mM tested with a potential of 30 mV
Catalog No. 50-002-67357 Supplier Medchemexpress LLC Supplier No. HYY0258S5MG
Benzocaine-d4 is the deuterium labeled Benzocaine Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na channel with an IC50 of 0 8 mM tested with a potential of 30 mV