Learn More
Medchemexpress LLC Benzocaine-(ethyl-d5) | 1219803-76-5 | 99.0% | 170.22 g/mol | C9H6D5NO2 | 10mg
SDP
Catalog No. 5000267890
Encompass_Preferred
Benzocaine-(ethyl-d5) is the deuterium labeled Benzocaine Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na channel with an IC50 of 0 8 mM tested with a potential of 30 mV
Catalog No. 50-002-67890 Supplier Medchemexpress LLC Supplier No. HYY0258S110MG
Benzocaine-(ethyl-d5) is the deuterium labeled Benzocaine Benzocaine shares a common receptor with all othe rLAs in the voltage-gated Na channel with an IC50 of 0 8 mM tested with a potential of 30 mV