Learn More
Medchemexpress LLC Benzocaine hydrochloride | 23239-88-5 | MFCD00012999 | 201.65 | C9H12ClNO2 | 500mg
SDP
Catalog No. 5000267356
Encompass_Preferred
Benzocaine (hydrochloride) is an orally active local agent that can suppress or relieve pain that acts on voltage-gated Na channels at a common receptor with an IC50 value of 0 8 mM at 30 mV Additionally Benzocaine (hydrochloride) non-competitively inhibits Ca-ATPase binding with Ca2 with an IC50 of 47 1 mM Benzocaine (hydrochloride) can be used in research within the field of neuromuscular regulation[1][2][3]
Catalog No. 50-002-67356 Supplier Medchemexpress LLC Supplier No. HYY0258A500MG
Benzocaine (hydrochloride) is an orally active local agent that can suppress or relieve pain that acts on voltage-gated Na channels at a common receptor with an IC50 value of 0 8 mM at 30 mV Additionally Benzocaine (hydrochloride) non-competitively inhibits Ca-ATPase binding with Ca2 with an IC50 of 47 1 mM Benzocaine (hydrochloride) can be used in research within the field of neuromuscular regulation[1][2][3]