Learn More
Medchemexpress LLC Benzofuran, 2,3-dihydro- | 496-16-2 | 99.5% | C8H8O | 25 G
SDP
Catalog No. 5000359191
Encompass_Preferred
Coumaran (2,3-Dihydrobenzofuran) acts as an AChE inhibitor and possesses antileishmanial activity. It may acquire antiparasitic capabilities through activation of macrophages and exert immunomodulatory activity. This compound can also be utilized as a biopesticide.
  • Active against promastigote and amastigote forms
  • Not significantly cytotoxic to macrophages or erythrocytes
  • Increases phagocyte and lysosomal activity
  • Increases nitrite (NO2-) level
  • Inhibits AChE
  • Acts as an endogenous metabolite
  • Derived from plants such as Verbenaceae Lantana camaraLinn.
Catalog No. 50-003-59191 Supplier Medchemexpress LLC Supplier No. 25GHY75247
Coumaran (2,3-Dihydrobenzofuran) acts as an AChE inhibitor and possesses antileishmanial activity. It may acquire antiparasitic capabilities through activation of macrophages and exert immunomodulatory activity. This compound can also be utilized as a biopesticide.
  • Active against promastigote and amastigote forms
  • Not significantly cytotoxic to macrophages or erythrocytes
  • Increases phagocyte and lysosomal activity
  • Increases nitrite (NO2-) level
  • Inhibits AChE
  • Acts as an endogenous metabolite
  • Derived from plants such as Verbenaceae Lantana camaraLinn.