Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzofuran-3-carbaldehyde | 4687-25-6 | 146.15 | 100 MG
SDP

Catalog No. 5000446068
Encompass_Preferred

Benzofuran-3-carbaldehyde is a bioactive chemical used for the synthesis of active compounds. This product is intended for research use only and is available as a light brown to orange solid.

  • Chemical formula C9H6O2
  • Molecular weight 146.15
  • Storage at 4°C under nitrogen
  • In solvent storage at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-46068 Supplier Medchemexpress LLC Supplier No. HYW004801100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzofuran-3-carbaldehyde is a bioactive chemical used for the synthesis of active compounds. This product is intended for research use only and is available as a light brown to orange solid.

  • Chemical formula C9H6O2
  • Molecular weight 146.15
  • Storage at 4°C under nitrogen
  • In solvent storage at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.