Learn More
Medchemexpress LLC Benzofuran-3-carbaldehyde | 4687-25-6 | 146.15 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW004801500MG
Encompass_Preferred
Benzofuran-3-carbaldehyde is a bioactive chemical used for the synthesis of active compounds. It is intended for research use only.
  • Used for synthesizing active compounds
  • Appears as a solid with a light brown to orange color
  • Exhibits high solubility in DMSO (100 mg/mL)
  • Supports multiple in vivo solubility protocols
Catalog No. 50-004-49086