Learn More
Medchemexpress LLC Benzohydroxamic acid | 495-18-1 | MFCD00002109 | 100.0% | 137.14 g/mol | C7H7NO2 | 100g
SDP
Catalog No. 5000267990
Encompass_Preferred
Benzohydroxamic acid (BHA) is a chitin deacetylase (CDA) inhibitor with significant antifungal activity The Ki values of BHA against the CDAs of Verticillium dahliae and Puccinia striiformis f sp tritici are 8 31 M and 9 83 M respectively Benzohydroxamic acid can restore the defense responses of infected host plants upregulate the expression of defense-related genes and reduce the growth and reproduction of fungi in plants Benzohydroxamic acid can be used in the research of the field of controlling agricultural fungal diseases such as various plant fungal diseases caused by Verticillium dahliae Puccinia striiformis and other fungi like cotton wilt and wheat stripe rust [1]
Catalog No. 50-002-67990 Supplier Medchemexpress LLC Supplier No. HYY1058100G
Add to Cart
Add to Cart
Benzohydroxamic acid (BHA) is a chitin deacetylase (CDA) inhibitor with significant antifungal activity The Ki values of BHA against the CDAs of Verticillium dahliae and Puccinia striiformis f sp tritici are 8 31 M and 9 83 M respectively Benzohydroxamic acid can restore the defense responses of infected host plants upregulate the expression of defense-related genes and reduce the growth and reproduction of fungi in plants Benzohydroxamic acid can be used in the research of the field of controlling agricultural fungal diseases such as various plant fungal diseases caused by Verticillium dahliae Puccinia striiformis and other fungi like cotton wilt and wheat stripe rust [1]