Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzohydroxamic Acid 98.0+%, TCI America™
SDP

Catalog No. B006125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006125G 25 g
B00615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B006125G Supplier TCI America Supplier No. B006125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 495-18-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00002109
InChI Key VDEUYMSGMPQMIK-UHFFFAOYSA-N
Synonym benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
PubChem CID 10313
IUPAC Name N-hydroxybenzamide
SMILES C1=CC=C(C=C1)C(=O)NO

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzohydroxamic Acid
RTECSNumber DF9650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.