Learn More
Medchemexpress LLC Benzoic-1,2,3,4,5,6-13C6 acid, 4-hydroxy-, butyl ester | 1416711-53-9 | 99.8% | 500 UG
SDP
Catalog No. 5000413522
Encompass_Preferred
Butylparaben-13C6 is the 13C labeled Butylparaben. Butylparaben is an organic compound that has proven to be a highly successful antimicrobial preservative in cosmetics, also used in medication suspensions, and as a flavoring additive in food. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13522 Supplier Medchemexpress LLC Supplier No. HYB1431S1500UG
Butylparaben-13C6 is the 13C labeled Butylparaben. Butylparaben is an organic compound that has proven to be a highly successful antimicrobial preservative in cosmetics, also used in medication suspensions, and as a flavoring additive in food. This compound is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS