Learn More
Medchemexpress LLC Benzoic-2,3,4,5-d4 acid, 6-(acetyloxy)- | 97781-16-3 | 97.7% | C9H4D4O4 | 10MG
SDP
Catalog No. 5000258386
Encompass_Preferred
Aspirin-d4 (acetylsalicylic acid-d4) is a deuterium-labeled form of aspirin used as an isotopically labeled internal standard and tracer for quantitative analytical techniques. Supplied as a solid with high reported purity, it is intended for analytical workflows that require reliable internal standards for accurate quantitation.
  • Deuterium-labeled acetylsalicylic acid for use as an internal standard.
  • High purity (97.7%) suitable for quantitative analysis.
  • Solid physical form for convenient handling and storage.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Available in small milligram pack sizes for analytical workflows.
Catalog No. 50-002-58386 Supplier Medchemexpress LLC Supplier No. HY14654S110MG
Add to Cart
Add to Cart
Aspirin-d4 (acetylsalicylic acid-d4) is a deuterium-labeled form of aspirin used as an isotopically labeled internal standard and tracer for quantitative analytical techniques. Supplied as a solid with high reported purity, it is intended for analytical workflows that require reliable internal standards for accurate quantitation.
  • Deuterium-labeled acetylsalicylic acid for use as an internal standard.
  • High purity (97.7%) suitable for quantitative analysis.
  • Solid physical form for convenient handling and storage.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Available in small milligram pack sizes for analytical workflows.