Learn More
Medchemexpress LLC Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, propyl ester | 1219802-67-1 | 184.23 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN2026S15MG
Encompass_Preferred
Propylparaben-d4 is the deuterium-labeled version of Propylparaben, an antimicrobial preservative naturally produced by plants and bacteria. It is commonly used in cosmetics, pharmaceuticals, and foods. Propylparaben has been shown to disrupt antral follicle growth and steroidogenic function by altering cell-cycle, apoptosis, and steroidogenesis pathways, and also decreases sperm number and motile activity in rats.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-15877