Learn More
Selleck Chemical LLC Benzoic Acid S4161-50mg
SDP
Catalog No. 503108386
Encompass_Preferred
Benzoic acid (FEMA 2131) is a colorless crystalline solid and a simple aromatic carboxylic acid used as a food preservative
Catalog No. 50-310-8386 Supplier Selleck Chemical LLC Supplier No. S416150MG
Benzoic acid (FEMA 2131) is a colorless crystalline solid and a simple aromatic carboxylic acid used as a food preservative