Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 2-[[(18S,21S,22R)-22-amino-21-hydroxy-18-(2-methylpropyl)-... | 1351169-27-1 | 97.9% | 917.14 g/mol | C51H72N4O11 | 5 MG
SDP

Catalog No. 5000263193
Encompass_Preferred

PROTAC RAR Degrader-1 is a proteolysis-targeting chimera designed to induce degradation of retinoic acid receptor family members by recruiting IAP E3 ligases. It is supplied as a solid research compound characterized by formula C51H72N4O11 and a molecular weight of 917.14 g/mol, intended for cellular studies of RAR/RXR biology.

  • Induces degradation of retinoic acid receptors.
  • Targets cIAP1 and RXRα.
  • Molecular formula C51H72N4O11; molecular weight 917.14 g/mol.
  • Purity 97.9%.
  • Appearance: solid.
  • Solubility: DMSO ≥ 80 mg/mL (87.23 mM).
  • Storage: -20°C under nitrogen; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-63193 Supplier Medchemexpress LLC Supplier No. HY1118445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC RAR Degrader-1 is a proteolysis-targeting chimera designed to induce degradation of retinoic acid receptor family members by recruiting IAP E3 ligases. It is supplied as a solid research compound characterized by formula C51H72N4O11 and a molecular weight of 917.14 g/mol, intended for cellular studies of RAR/RXR biology.

  • Induces degradation of retinoic acid receptors.
  • Targets cIAP1 and RXRα.
  • Molecular formula C51H72N4O11; molecular weight 917.14 g/mol.
  • Purity 97.9%.
  • Appearance: solid.
  • Solubility: DMSO ≥ 80 mg/mL (87.23 mM).
  • Storage: -20°C under nitrogen; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.