Learn More
Medchemexpress LLC Benzoic acid, 2-[(2-carboxyphenyl)amino]-4-chloro-, disodium salt | 64808-48-6 | MFCD00941422 | 99.9% | 335.65 g·mol⁻¹ | C14H8ClNNa2O4 | 5 MG
SDP
Catalog No. 5000240620
Encompass_Preferred
Lobenzarit disodium is the disodium salt form of lobenzarit (CCA), a small-molecule anti-arthritic and antioxidant agent supplied as a purified reagent for preclinical in vitro and in vivo research. It is used to study anti-inflammatory effects, modulation of reactive oxygen species, and related pharmacological activities.
  • Anti-arthritic and antioxidant small molecule for preclinical research.
  • High purity reagent (>99.9%).
  • Disodium salt form for improved aqueous handling compared with the free acid.
  • Suitable for in vitro and in vivo anti-inflammatory and antioxidant assays.
  • Molecular formula C14H8ClNNa2O4; molecular weight 335.65 g·mol⁻¹.
Catalog No. 50-002-40620 Supplier Medchemexpress LLC Supplier No. HY1073605MG
Lobenzarit disodium is the disodium salt form of lobenzarit (CCA), a small-molecule anti-arthritic and antioxidant agent supplied as a purified reagent for preclinical in vitro and in vivo research. It is used to study anti-inflammatory effects, modulation of reactive oxygen species, and related pharmacological activities.
  • Anti-arthritic and antioxidant small molecule for preclinical research.
  • High purity reagent (>99.9%).
  • Disodium salt form for improved aqueous handling compared with the free acid.
  • Suitable for in vitro and in vivo anti-inflammatory and antioxidant assays.
  • Molecular formula C14H8ClNNa2O4; molecular weight 335.65 g·mol⁻¹.