Learn More
Medchemexpress LLC Benzoic acid, 2-[4-[2-(1-piperidinyl)ethoxy]benzoyl]-, methyl ester, hydrochloride (1:1) | 1248-42-6 | 99.9% | 403.90 | 25 MG
SDP
Catalog No. 5000345537
Encompass_Preferred
Pitofenone hydrochloride is a spasmolytic compound that inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and electric eel with Kis of 36 and 45 μM, respectively. It also exhibits anticonvulsant activity when combined with diclofenac and fenpivirine.
  • Spasmolytic compound
  • Inhibits acetylcholinesterase (AChE) activity
  • Anticonvulsant activity in combination with diclofenac and fenpivirine
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-45537 Supplier Medchemexpress LLC Supplier No. 25MGHY110389
Pitofenone hydrochloride is a spasmolytic compound that inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and electric eel with Kis of 36 and 45 μM, respectively. It also exhibits anticonvulsant activity when combined with diclofenac and fenpivirine.
  • Spasmolytic compound
  • Inhibits acetylcholinesterase (AChE) activity
  • Anticonvulsant activity in combination with diclofenac and fenpivirine
  • Appearance: Solid
  • Color: White to off-white